Hydrostatic paradox
Hydrostatic paradox
Katalog # 765-0138
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Shows that the bottom pressure in a liquid depends only on the height and not the shape of the container. The fluid pressure is transferred via a rubber diaphragm to an adjustable hand. Comes with four containers of different shapes. Dimensions.
- Height: 290 mm
- Bottom plate: 235 × 130 mm
Specifikationer
- Beskrivelse:Hydrostatic paradox