Call for surface tension test
Call for surface tension test
Katalog # 765-0136
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Aluminum ring with suspension in three points. Used together with a dynamometer to determine the surface tension in liquids.
- Diameter: 65 mm
Specifikationer
- Beskrivelse:Call for surface tension test