Orders LinkContactUsLinkComponent
Buoyancy measuring apparatus
Buoyancy measuring apparatus
Buoyancy measuring apparatus
Katalog # 765-0139
Marketsource
Leverandør:   CORNELSEN EXPERIMENTA
Buoyancy measuring apparatus
Katalog # 765-0139
Leverandør:  CORNELSEN EXPERIMENTA
Marketsource
Buoyancy measuring apparatus
Buoyancy measuring apparatus
Buoyancy measuring apparatus
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Equipment to determine the buoyancy of different materials. It consists of three balls - wood, styrofoam and plastic, a pulley, and twine. The balls incorporate a hook for attachment of twine. Stand not included.
  • Ball Ø: 80 mm
Specifikationer
  • Beskrivelse:
    Buoyancy measuring apparatus