Buoyancy measuring apparatus
Buoyancy measuring apparatus
Katalog # 765-0139
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Equipment to determine the buoyancy of different materials. It consists of three balls - wood, styrofoam and plastic, a pulley, and twine. The balls incorporate a hook for attachment of twine. Stand not included.
- Ball Ø: 80 mm
Specifikationer
- Beskrivelse:Buoyancy measuring apparatus