Recipient
Recipient
765-0146
Marketsource
Recipient with opening at the top. With the help of a rubber plug with electrical connection bushings, for example, a bell can be hung up in ordinary laboratory cables.
- Height, inv: 300 mm
- Diameter, inner: 200 mm
- Beskrivelse:Recipient