Daniell element
Daniell element
Catalog # 765-0199
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Used in electrochemistry. Voltage, max 1 V. The unit is divisible and consists of glass beaker, cylindrical zinc electrode, plastic beaker, cylindrical copper electrode and crocodile clamps. Fill with copper sulphate (10%) or zinc sulphate (10%). A light-rotating motor with a propeller is suitable as a voltage indicator.
Specifications
- Beskrivelse:Daniell element