Bunsen element
Bunsen element
765-0201
Marketsource
Used in electrochemistry. Voltage, max 2 V. The unit is divisible and consists of glass beaker, cylindrical zinc electrode, plastic beaker, carbon electrode, crocodile clamps and lid. Fill with nitric acid (65%) or sulfuric acid (8%). A light-rotating motor with a propeller is suitable as a voltage indicator.
- Beskrivelse:Bunsen element