Melting gutter
Melting gutter
Katalog # 765-0206
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Circular gutter made of aluminum sheet with insulating handle. Fits U-core 765-1077 and was used, among other things, when demonstrating the principle of induction furnaces.
- Gutter: Diam, outer × inner: 95 × 70 mm
Handle: Length × Diam: 100 × 10 mm
Specifikationer
- Beskrivelse:Melting gutter