Electrostatic double pendulum
Electrostatic double pendulum
Katalog # 765-0180
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Used to detect electric charge. Also suitable for use with belt generator and Faraday's cage. Equipped with connection for 4 mm banana plug.
- Dimensions: total height 150 mm, cylinders: 10 × 10 mm
Specifikationer
- Beskrivelse:Electrostatic double pendulum