Plastic fibers
Plastic fibers
Katalog # 765-0193
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For demonstration of electric field lines and electrostatic charge.
- 30 g in a plastic bag
Specifikationer
- Beskrivelse:Plastic fibers