Electroscope
Electroscope type Braun
Katalog # 765-0181
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For electrostatic experiments. Consists of insulated aluminum holder with pointed bearing, removable capacitor plate, metal ring with rod and 4 mm earthing contact.
- Dimension: metal ring diam. 200 mm, rod diam. 10 mm
Specifikationer
- Beskrivelse:Electroscope type Braun