Electrometer
Electrometer
Catalog # 765-0182
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Instrument for comparing charge amounts. When charging, a hand is repelled from the jumper it is hung in. Mounted on a plate.
Specifications
- Beskrivelse:Electrometer