Needle holder with pin
Needle holder with pin
Katalog # 765-0191
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For attaching an electric spinner to the ball generator's ball. Equipped with 4 mm banana plug.
Specifikationer
- Beskrivelse:Needle holder with pin
- Cat. No.:765-0191