Plexiglas rod
Plexiglas rod
Katalog # 765-0184
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Used in electrostatic experiments. Equipped with holes, 4 mm, at one end for connection of conductor ball or capacitor plate.
- Dimensions: 150 × 15 mm
Specifikationer
- Beskrivelse:Plexiglas rod