Rod
Rod
Katalog # 765-0198
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
Insulated rod with 3 connection terminals for wire up to 2 mm and 4 mm contact sleeve.
- Dimensions: rod diam 10 × 55 mm, height 120 mm
Specifikationer
- Beskrivelse:Rod