Insulating rod
Insulating rod
Katalog # 765-0197
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
High voltage porcelain insulator. The high-voltage side is equipped with two radial and one axial holes for 4 mm banana pins and a slot with a clamping screw for plates with a maximum thickness of 4 mm. The grounded side has a radial hole for 4 mm banana pins. Insulation voltage: max 25 kV.
- Dimensions: 10 (dia) × 180 mm
Specifikationer
- Beskrivelse:Insulating rod