Conductor bottle
Conductor bottle
Katalog # 765-0189
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For storage of electric charge. Consists of a glass cylinder with metal foil and a conductor ball on a rod.
- Capacitance: about 1000 pF
- Dimensions: 260 × 80 mm (diam)
Specifikationer
- Beskrivelse:Conductor bottle