Electrostatic motor
Electrostatic motor
Catalog # 765-0186
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Consists of a plastic cylinder placed on an insulating rod between two tip electrodes. When connecting a high-voltage source, a charge transport is obtained which causes the cylinder to rotate.
- The following are included: rotor with bearings, rod with needle, tip electrodes with 4 mm contact pins (4 pcs.), Insulating rods with 4 mm contact sleeves (2 pcs.). Suitable accessory: influence machine
Specifications
- Beskrivelse:Electrostatic motor