Orders LinkContactUsLinkComponent
Modular diode laser
Modular diode laser
Leverandør:   KVANT LIFE SCIENCE
Modular diode laser
Leverandør:  KVANT LIFE SCIENCE
Certifikater
Modular diode laser
Modular diode laser
1 tilgængelige muligheder nedenfor
Product Details & Documents
The advantages of a diode laser compared to a HeNe laser are mainly small format, low power consumption and long service life, as well as the possibility of electronic modulation. The laser is modular in the range 100 Hz 20 MHz. Dimensions.
  • Elastic in width and thus easy to apply
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • 765-1004
    Modular diode laser, 635 nm, 3 mW
    Modular diode laser, 635 nm, 3 mW
    Discontinued
  • 765-1018
    Modular diode laser, 635 nm, 1 mW
    Modular diode laser, 635 nm, 1 mW