Transport trolley
Transport trolley
Katalog # 765-0998
Leverandør: JOHANSSON INDUSTRI AB HJALMA
Gå til:
Product Details & Documents
Suitable for transport of appliances, instruments, counters, etc. Made of galvanized galvanized steel pipe and equipped with shelves of Perstorp plate and ball wheels.
- Length: 780 mm
- Shelves: 760 × 430 mm
- Handlebar height: 960 mm
Specifikationer
- Beskrivelse:Transport trolley