Bottle Carrier
Bottle rack
Katalog # 766-0169
Marketsource
Leverandør: GR NNA PLASTEMBALLAGE AB
Gå til:
Product Details & Documents
Plastic.
- Stackable
- Bottle rack with carrying handle
- For 4×500 ml and 4×250 ml bottles
Colour: Blue
Specifikationer
- Beskrivelse:Bottle rack