Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, 3,3′,5,5′-Tetramethylbenzidine

Supelco® Organic Reference Standard, 3,3′,5,5′-Tetramethylbenzidine

3,3′,5,5′-Tetramethylbenzidine, Standard for GC, Glass bottle for solids, 250 mg
Katalog # 09743-250MG
Leverandør:   Merck
Supelco® Organic Reference Standard, 3,3′,5,5′-Tetramethylbenzidine
Katalog # 09743-250MG
Leverandør:  Merck
CAS nummer:  
Supelco® Organic Reference Standard, 3,3′,5,5′-Tetramethylbenzidine
Supelco® Organic Reference Standard, 3,3′,5,5′-Tetramethylbenzidine

3,3′,5,5′-Tetramethylbenzidine, Standard for GC, Glass bottle for solids, 250 mg

CAS nummer:  
Price
300,00 kr
Log ind for tilgængelighed
1 * 250 mg
Product Details & Documents

Caution: P202 - P264 - P273 - P280 - P301 + P312 - P308 + P313

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 mg
  • Standard:
    Standard for GC
  • Beskrivelse:
    3,3′,5,5′-Tetramethylbenzidine
  • Cat. No.:
    09743-250MG
  • M-Clarity quality level:
    MQ100
  • CAS-nr:
    54827-17-7
  • Molekylvægt:
    240,34
  • MDL No.:
    MFCD00007748
  • Fareklassebeskrivelse:
    H302 - H351 - H413
  • Flammepunkt:
    Not applicable
  • Opbevaring betingelser:
    2...30 °C
  • Melting point:
    166...171 °C; 168...171 °C (lit.)
  • Synonym(s):
    BM blue; Sure Blue TMB; TMB; TMB Blotting Plus; TMB substrate
  • Shipping conditions:
    STD
  • Formel:
    [-C₆H₂(CH₃)₂-4-NH₂]₂

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.