Orders LinkContactUsLinkComponent
Neutral rød
Glass bottle for solids, 25 g
Katalog # SERA30305.01
Marketsource
Leverandør:   Serva
Neutral rød
Katalog # SERA30305.01
Leverandør:  Serva
Marketsource
CAS nummer:  553-24-2
Neutral rød
Glass bottle for solids, 25 g
CAS nummer:  553-24-2
Price
597,41 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS nummer:553-24-2
  • Formel:C₁₅H₁₇ClN₄
  • MDL nummer:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:12,06571
  • Smeltende Pt:290 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-035-8
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g
Specifikationer