Orders LinkContactUsLinkComponent
Galdesyre 97%
Irritant
Galdesyre 97%
Certificates
Galdesyre 97%
3 Options Available Below
Product Details & Documents
Synonyms:  3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
  • CAS Number:81-25-4
  • Formula:C₂₄H₄₀O₅
  • MDL Number:MFCD00003672
  • Molecular Weight:408.58 g/mol
  • Merck Index:13,02223
  • Boiling Pt:545 °C (1013 hPa)
  • Melting Pt:197…202 °C
  • Storage Temperature:Stue temperatur
  • EINECS:201-337-8
Specifications for Product Family
Specification Test Results
  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    197 to 202 °C
  • Loss on drying:
    ≤1 % (135 °C)
  • Heavy metals:
    ≤20 ppm
  • Sulfated ash:
    ≤0.1 %
  • TLC:
    ≥96.0 %
  • Specific optical rotation:
    (20 °C, 589 nm) (c=2, ethanol) > +35°
  • Titration with NaOH:
    ≥96.0 %
  • UV:
    at 370 nm A: ≤0.004
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Chloride (Cl):
    ≤0.0005 %
  • Sulfate (SO4):
    ≤0.0005 %
  • Cadmium (Cd):
    ≤0.000005 %
  • Copper (Cu):
    ≤0.000002 %
  • Ammonia (NH3):
    ≤0.002 %
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 327,00 kr
    327,00 kr
    1 * 25 g
       327,00 kr
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 699,00 kr
    699,00 kr
    1 * 100 g
       699,00 kr
  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2.890,00 kr
    2.890,00 kr
    1 * 500 g
       2.890,00 kr