General purpose thermometer
Soil thermometer, 0…+80 °C
Katalog # 763-0411
Marketsource
Leverandør: MOLLER THERM
Gå til:
Product Details & Documents
Housing made of plastic. Gradation −5 to 65 °C.
Specifikationer
- Beskrivelse:Soil thermometer
- Område:0…+80 °C