Demonstration range for oh
Catalog # 765-0116
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Product Details & Documents
Demonstration model of a caliper. Made of transparent plastic. Equipped with Nonies scale and rubber feet. Intended for OHapparat. Can also be used independently.
- Dimensions: 200 × 105 mm