Cell building
Cell structure model, SOMSO-Plast®
Katalog # 763-0452
Leverandør: SOMMER MARCUS
Gå til:
Product Details & Documents
The 10000x magnification model shows the structure of an animal cell. Cell plasma with mitochondria, centrioles, endoplasmic reticulum with and without ribosomes. Cell nucleus with nuclear membranes, pores, nucleols, chromatin.
Leveringsinformation: 22 cm high cell model mounted on a stand with a base.
Specifikationer
- Beskrivelse:Cell structure model
- Materiale:SOMSO-Plast®