Platinum loop with glass handle
Platinum loop with glass handle
765-1396
: LABGLAS
Total length approx. 200 mm, glass shaft 150×5 mm. Thread diameter 0,5 mm, inner diameter of the loop 1,5 mm.
- Beskrivelse:Platinum loop with glass handle