Orders LinkContactUsLinkComponent
Electrical energy sources
Electrical energy sources, experimental kit
Katalog # 765-0109
Marketsource
Leverandør:   CORNELSEN EXPERIMENTA
Electrical energy sources
Katalog # 765-0109
Leverandør:  CORNELSEN EXPERIMENTA
Marketsource
Electrical energy sources
Electrical energy sources
Electrical energy sources, experimental kit
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The kit contains about 15 different components with which a number of experiments can be performed. The components are designed so that students can quickly and easily perform their laboratory work. The kit can show how the following forms of energy are generated:
  • Thermal energy (thermocouple)
  • Radiant energy (solar cell)
  • Chemical energy (galvanic element)
  • Mechanical energy (induction)
To demonstrate that electrical energy is generated, the units can be connected to an included micromotor with propeller. Detailed instructions for use included. The kit contains i.a. the following components: Micro motor, 60 mV / 30 mA, Solar cell, 0.5 V / 300 mA, Flashlight, Electrolysis bath, Copper and zinc electrodes, Paper strips, Citric acid, Plastic tube with two rings, Rod magnet, Copper and constant wire.
Storage box dimensions: 300 × 170 × 40 mm
Specifikationer
  • Beskrivelse:
    Electrical energy sources, experimental kit