Basic Gas Chromatography
Basic gas chromatography
Katalog # 766-0182
Leverandør: WILEY JOHN SONS
Gå til:
Product Details & Documents
Basic book on gas chromatography. Principles, calculations in qualitative and quantitative analyzes. How different detectors such as TCD, FID etc. works. Structure and properties of different stationary phases and support materials. Suitable for you who have or are considering obtaining our school chromatograph GC82 12.
Specifikationer
- Beskrivelse:Basic gas chromatography