Laser communication
Laser communication
Katalog # 765-1013
Leverandør: KVANT LIFE SCIENCE
Gå til:
Product Details & Documents
Equipment for laser sound transmission experiments. The included microphone, tape recorder or function generator is used as the sound source. The rate consists of.
- Modular laser (635 nm / 1mW)
Specifikationer
- Beskrivelse:Laser communication