Orders LinkContactUsLinkComponent
6-ROX alkyne
6-ROX alkyne
5 mg
Catalog # AATB495
Supplier:   AAT Bioquest
6-ROX alkyne
Catalog # AATB495
Supplier:  AAT Bioquest
CAS Number:  
6-ROX alkyne
6-ROX alkyne
5 mg
CAS Number:  
Price
1.770,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents
6-ROX alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Caution: For research use only.
  • Molecular Weight:571.66 g/mol
Specifications
Specifications
  • Pakn:
    5 mg