Orders LinkContactUsLinkComponent
iFluor® 568, Tyramide
200
Katalog # AATB45106
Leverandør:   AAT Bioquest
iFluor® 568, Tyramide
Katalog # AATB45106
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 568, Tyramide
iFluor® 568, Tyramide
200
CAS nummer:  
Price
3.360,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
 : For research use only.
Specifikationer
Specifikationer
  • Pakn:
    200
Specifikationer