Orders LinkContactUsLinkComponent
iFluor® 555, Azide
iFluor® 555, Azide
Plastic bottle, 1 mg
Katalog # AATB1093
Irriterende
Leverandør:   AAT Bioquest
iFluor® 555, Azide
Katalog # AATB1093
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 555, Azide
iFluor® 555, Azide
Plastic bottle, 1 mg
CAS nummer:  
Price
2.350,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • ADR:5,II
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • Pakn:
    1 mg