Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
iFluor® 405, Amine

iFluor® 405, Amine

Plastic bottle, 1 mg
Katalog # AATB1071
Irriterende
Leverandør:   AAT Bioquest
iFluor® 405, Amine
Katalog # AATB1071
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 405, Amine
iFluor® 405, Amine

Plastic bottle, 1 mg

CAS nummer:  
Price
2.350,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • ADR:5,II
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.