Cell Meter™ VX450 Fixable Viability Dye
Cell Meter™ VX450 fixable cell stain, 200 Tests, 200 Tests
Katalog # AATB22540
Leverandør: AAT Bioquest
Gå til:
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
- Cell-impermeable
- Amine- and thiol-reactive
- High stability
- Live/dead cell labeling dye
- Fixable
Advarsel: For research use only.
Specifikationer
Specifikationer
- Pakn:200 Tests
- Beskrivelse:Cell Meter™ VX450 fixable cell stain
- Test pr. sæt:200 Tests