Orders LinkContactUsLinkComponent
Cell Meter™ RX700 Fixable Viability Dye
Cell Meter™ RX700 Fixable Viability Dye
Cell Meter™ RX700 fixable cell stain, 200 Tests, 200 Tests
Katalog # AATB22532
Leverandør:   AAT Bioquest
Cell Meter™ RX700 Fixable Viability Dye
Katalog # AATB22532
Leverandør:  AAT Bioquest
CAS nummer:  
Cell Meter™ RX700 Fixable Viability Dye
Cell Meter™ RX700 Fixable Viability Dye
Cell Meter™ RX700 fixable cell stain, 200 Tests, 200 Tests
CAS nummer:  
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-impermeable
  • Amine- and thiol-reactive
  • High stability
  • Live/dead cell labeling dye
  • Fixable
Advarsel: For research use only.
Specifikationer
Specifikationer
  • Pakn:
    200 Tests
  • Beskrivelse:
    Cell Meter™ RX700 fixable cell stain
  • Test pr. sæt:
    200 Tests
Specifikationer