Orders LinkContactUsLinkComponent
PUR-1
PUR-1
PUR-1, 1 mg
  AATB17560
PUR-1
  AATB17560
 :  
PUR-1
PUR-1
PUR-1, 1 mg
 :  
1.170,00 kr
1 * 1 mg
PUR-1 is essentially non-fluorescent in the absence of nucleic acids.
  • Useful For Multicolor Imaging
  • Bright Fluorescence
  • 40% Fluorescence Quantum Yield
Caution: For research use only.
  • Pakn:
    1 mg
  • Beskrivelse:
    PUR-1