PUR-1
PUR-1, 1 mg
AATB17560
PUR-1 is essentially non-fluorescent in the absence of nucleic acids.
- Useful For Multicolor Imaging
- Bright Fluorescence
- 40% Fluorescence Quantum Yield
Caution: For research use only.
- Pakn:1 mg
- Beskrivelse:PUR-1