Heat radiation
Heat radiation
Katalog # 765-0177
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For experiments with absorption of heat radiation. Consists of a black and a white E-flask and two thermometers (0 to 50 °C).
- Comes with insulating cork plate
Specifikationer
- Beskrivelse:Heat radiation