Cell Meter™ Live Cell Caspase 6 Binding Assay Kit *Green Fluorescence*
Catalog # AATB20113
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pakn:25 Tests
- Beskrivelse:Cell Meter™ Live cell Caspase 6 binding assay kit *Green fluorescence*
- Test pr. sæt:25 Tests
Specifications
About this item
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms
Caution: For research use only.