Cell Meter™ Intracellular NADH/NADPH Fluorescence Imaging Kit *Deep Red Fluorescence*
Katalog # AATB15295
Leverandør: AAT BIOQUEST
CAS nummer:
Specifikationer
- Pakn:100 Tests
- Beskrivelse:Cell Meter™ Intracellular NADH/NADPH fluorescence imaging kit *deep red fluorescence*
- Test pr. sæt:100 Tests
Specifikationer
Om denne vare
The detection of intracellular dihydronicotinamide adenine dinucleotide NADH and its phosphate ester NADPH is important for disease diagnostics and drug discovery.
- High sensitivity
- Bright fluorescence
- Suitable for live cell imaging
Caution: For research use only.