Orders LinkContactUsLinkComponent
MitoLite™ NIR FX690
MitoLite™ NIR FX690
MitoLite™ NIR FX690, 500 Tests, 500 Tests
Katalog # AATB22690
Leverandør:   AAT Bioquest
MitoLite™ NIR FX690
Katalog # AATB22690
Leverandør:  AAT Bioquest
CAS nummer:  
MitoLite™ NIR FX690
MitoLite™ NIR FX690
MitoLite™ NIR FX690, 500 Tests, 500 Tests
CAS nummer:  
Price
1.170,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Mitochondria are membrane-enclosed organelles found in most eukaryotic cells.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Advarsel: For research use only.
Specifikationer
Specifikationer
  • Pakn:
    500 Tests
  • Beskrivelse:
    MitoLite™ NIR FX690
  • Test pr. sæt:
    500 Tests
  • Cat. No.:
    AATB22690
  • Renhed:
    90%