MitoLite™ Red FX600
MitoLite™ Red, 500 Tests, 500 Tests
AATB22677
MitoLite™ Red is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
: For research use only.
- Pakn:500 Tests
- Beskrivelse:MitoLite™ Red
- Test pr. sæt:500 Tests
- Cat. No.:AATB22677
- Renhed:95%