Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Biocytin ≥90%

Biocytin ≥90%

Plastic bottle, 100 mg
Katalog # AATB3080
Irriterende
Leverandør:   AAT Bioquest
Biocytin ≥90%
Katalog # AATB3080
Leverandør:  AAT Bioquest
CAS nummer:  576-19-2
Biocytin ≥90%
Biocytin ≥90%

Plastic bottle, 100 mg

CAS nummer:  576-19-2
Price
1.770,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Biocytin is invaluable as a rapidly transporting, sensitive neuronal tracer requiring little permeabilizing agents.

Synonymer:  ε-Biotinoyl-L-lysine, H-Lys(biotinyl)-OH
  • CAS nummer:576-19-2
  • Formel:C₁₆H₂₈N₄O₄S
  • MDL nummer:MFCD00077319
  • Molecular Weight:372.484 g/mol
  • ADR:5,II
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.