Orders LinkContactUsLinkComponent
iFluor® 810, Acid
iFluor® 810, Acid
Plastic bottle, 1 mg
Katalog # AATB1385
Irriterende
Leverandør:   AAT Bioquest
iFluor® 810, Acid
Katalog # AATB1385
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 810, Acid
iFluor® 810, Acid
Plastic bottle, 1 mg
CAS nummer:  
Price
kr2,350.00
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • ADR:5,II
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • Pakn:
    1 mg
Specifikationer