Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
iFluor® 555 fluorescent probe

iFluor® 555 fluorescent probe

Plastic bottle, 1 mg
Katalog # AATB1007
Irriterende
Leverandør:   AAT Bioquest
iFluor® 555 fluorescent probe
Katalog # AATB1007
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 555 fluorescent probe
iFluor® 555 fluorescent probe

Plastic bottle, 1 mg

CAS nummer:  
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.

  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal
  • ADR:5,II
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.