Color mix, complementary batch
Complementary kit for color mixing
Katalog # 765-1314
Marketsource
Leverandør: PHYWE
Gå til:
Product Details & Documents
Supplementary kit for table optics 765-1317. Content:
- 2 mirrors
- 1 column
- 3 filters for additive color mixing
- 3 filters for subtractive color mixing
Specifikationer
- Beskrivelse:Complementary kit for color mixing