Picture optics, completion kit
Supplementary kit with lenses
Katalog # 765-1009
Leverandør: KVANT LIFE SCIENCE
Gå til:
Product Details & Documents
New optical models for laser optics (15 parts). Consists of.
- concave lenses
- plank concave lenses
- convex lenses
- planar convex lenses
- equilateral prism
- rectangular prisms
- mirrors
- condenser lenses
- cubic beam splitters
Specifikationer
- Beskrivelse:Supplementary kit with lenses