UV screen
UV screen
Katalog # 765-0161
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
To display ultraviolet radiation. Half of the plate is coated with a fluorescent substance that emits light during UV radiation.
Dimensions: 150 × 100 mm
Specifikationer
- Beskrivelse:UV screen