Light energy
Light energy
Catalog # 765-0178
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Apparatus for examining the conversion from electrical energy to light and heat energy. Consists of a plastic container, lid with light bulb (6 V / 5 A) in base and a resistor. The container is filled with water and the temperature change is measured. Equipped with connections for 4 mm banana plug.
- Dimensions: 120 × 70 mm
Specifications
- Beskrivelse:Light energy