Thermocouple
Thermocouple
Katalog # 765-0212
Marketsource
Leverandør: CORNELSEN EXPERIMENTA
Gå til:
Product Details & Documents
For experiments with thermoelectric effects. Consists of brazed copper and constant wire. Equipped with connections for 4 mm banana connectors. Mounted on plate with rod, 10 mm.
Specifikationer
- Beskrivelse:Thermocouple